Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Phenylpivalamide | 6625-74-7 | 100.0% | 177.24 g/mol | C11H15NO | 25 G
SDP

Catalog No. 5000209160
Encompass_Preferred

N-Phenylpivalamide (Pivalanilide) is a high-purity small-molecule building block used as an intermediate in organic synthesis and medicinal chemistry research. It is supplied as a white to off-white solid with long-term stability under recommended storage conditions.

  • High-purity building block suitable for synthesis applications.
  • White to off-white solid formulation for ease of handling.
  • CAS number 6625-74-7 for unambiguous identification.
  • Purity suitable for research use: 99.98% (typical).
  • Pack size available: 25 G.
  • Storage stable at room temperature for extended shelf life.

Catalog No. 50-002-09160 Supplier Medchemexpress LLC Supplier No. 25GHYW043105
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Phenylpivalamide (Pivalanilide) is a high-purity small-molecule building block used as an intermediate in organic synthesis and medicinal chemistry research. It is supplied as a white to off-white solid with long-term stability under recommended storage conditions.

  • High-purity building block suitable for synthesis applications.
  • White to off-white solid formulation for ease of handling.
  • CAS number 6625-74-7 for unambiguous identification.
  • Purity suitable for research use: 99.98% (typical).
  • Pack size available: 25 G.
  • Storage stable at room temperature for extended shelf life.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.