Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-phenylpivalamide | 6625-74-7 | MFCD00043691 | 100.0% | 177.24 | C11H15NO | 5 G
SDP

Catalog No. 5000210170
Encompass_Preferred

N-Phenylpivalamide is an organic small-molecule building block used in the synthesis of biologically active compounds. It is represented by the formula C11H15NO and has a reported CAS number 6625-74-7 and high reported purity.

  • small-molecule building block for synthesis of biologically active compounds.
  • chemical formula C11H15NO.
  • molecular weight approximately 177.24 g/mol.
  • reported high purity suitable for research applications.
  • available in small-scale packages for laboratory use.

Catalog No. 50-002-10170 Supplier Medchemexpress LLC Supplier No. HYW0431055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Phenylpivalamide is an organic small-molecule building block used in the synthesis of biologically active compounds. It is represented by the formula C11H15NO and has a reported CAS number 6625-74-7 and high reported purity.

  • small-molecule building block for synthesis of biologically active compounds.
  • chemical formula C11H15NO.
  • molecular weight approximately 177.24 g/mol.
  • reported high purity suitable for research applications.
  • available in small-scale packages for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.