Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Phenylseleno)phthalimide, 98%, Thermo Scientific™

Catalog No. AC225220050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225220050 5 g
AC225220010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC225220050 Supplier Thermo Scientific Chemicals Supplier No. 225220050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71098-88-9
Molecular Formula C14H9NO2Se
Molecular Weight (g/mol) 302.202
InChI Key TYZFQSLJTWPSDS-UHFFFAOYSA-N
Synonym n-phenylseleno phthalimide, 1h-isoindole-1,3 2h-dione, 2-phenylseleno, 2-phenylselanyl isoindole-1,3-dione, npsp, acmc-20akaa, upcmld00wv-69, n-phenylseleno phthalimide, technical grade, 2-phenylselenamethyl benzo c azoline-1,3-dione, 2-phenylselanyl-1h-isoindole-1,3 2h-dione #
PubChem CID 590397
IUPAC Name 2-phenylselanylisoindole-1,3-dione
SMILES C1=CC=C(C=C1)[Se]N2C(=O)C3=CC=CC=C3C2=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 178°C to 183°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Fieser 09,366; 10,312; 11,417; 12,390; 17,283; 01,107
Formula Weight 302.19
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material N-(Phenylseleno)phthalimide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.