Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Phenylurea

Catalog No. P36959500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P36959500G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P36959500G Supplier Sigma Aldrich Supplier No. P36959500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-phenylpyrazole undergoes cyclometallation with rhodium trichloride to yield rac-di(++-chloro)tetrakis[2-(pyrazol-1-yl)phenyl-c1,n2α]dirhodium. the activation of the c-h bond of 1-phenylpyrazole autocatalyzed by the co-product hoac was studied.1-phenylpyrazole has been used:
  • In the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole
  • As cyclometallated ligand in the preparation of new heteroleptic iridium(iii) complexes

Specifications

CAS 64-10-8
Melting Point 145°C to 147°C (lit.)
Molecular Formula C7H8N2O
Linear Formula C6H5NHCONH2
MDL Number MFCD00007944
Quantity 5 g
Synonym Phenylcarbamide
Molecular Weight (g/mol) 136.15
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.