Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Phthaloyl-β-alanine | 3339-73-9 | 219.20 | 1 G
SDP

Catalog No. 5000445908
Encompass_Preferred

N-Phthaloyl-β-alanine (hapten FP) is an FX-type hapten characterized by the absence of a CH2S moiety. This compound can be effectively coupled to OVA through the mixed anhydride method and exhibits various heterologies, making it a versatile tool for specific research applications. This product is intended for research use only.

  • FX-type hapten
  • Coupling to OVA via mixed anhydride method
  • Features increased heterologies
  • White to off-white solid appearance
  • Purity of 99.2%
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-45908 Supplier Medchemexpress LLC Supplier No. HYW0082341G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Phthaloyl-β-alanine (hapten FP) is an FX-type hapten characterized by the absence of a CH2S moiety. This compound can be effectively coupled to OVA through the mixed anhydride method and exhibits various heterologies, making it a versatile tool for specific research applications. This product is intended for research use only.

  • FX-type hapten
  • Coupling to OVA via mixed anhydride method
  • Features increased heterologies
  • White to off-white solid appearance
  • Purity of 99.2%
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.