Learn More
Medchemexpress LLC N-Phthaloyl-β-alanine | 3339-73-9 | MFCD00023096 | 219.20 | 25 G
SDP
Catalog No. 5000448180
Encompass_Preferred
N-Phthaloyl-β-alanine (hapten FP) is an FX-type hapten that lacks a CH2S moiety. It can be coupled to OVA using the mixed anhydride method and exhibits more heterologies.
  • FX-type hapten without a CH2S moiety
  • Can be coupled to OVA using the mixed anhydride method
  • Has more heterologies
Catalog No. 50-004-48180 Supplier Medchemexpress LLC Supplier No. 25GHYW008234
N-Phthaloyl-β-alanine (hapten FP) is an FX-type hapten that lacks a CH2S moiety. It can be coupled to OVA using the mixed anhydride method and exhibits more heterologies.
  • FX-type hapten without a CH2S moiety
  • Can be coupled to OVA using the mixed anhydride method
  • Has more heterologies