Learn More
Selleck Chemical LLC N-piperidine Ibrutinib HCl E2201-5MG
SDP
Catalog No. 5000039170
Encompass_Preferred
N-piperidine Ibrutinib hydrochloride a reversible Ibrutinib derivative is a potent BTK inhibitor with IC50s of 51 0 and 30 7 nM for WT BTK and C481S BTK respectively
Catalog No. 50-000-39170 Supplier Selleck Chemical LLC Supplier No. E22015MG
N-piperidine Ibrutinib hydrochloride a reversible Ibrutinib derivative is a potent BTK inhibitor with IC50s of 51 0 and 30 7 nM for WT BTK and C481S BTK respectively