Learn More
Medchemexpress LLC N-piperidine Ibrutinib hydrochloride | 2231747-18-3 | 98.1% | 1 ML
SDP
Catalog No. 5000338989
Encompass_Preferred
N-piperidine Ibrutinib hydrochloride is a reversible Ibrutinib derivative and a potent BTK inhibitor. It demonstrates high efficacy with IC50s of 51.0 nM for WT BTK and 30.7 nM for C481S BTK. This compound serves as a valuable BTK ligand in the synthesis of various PROTACs, including potent PROTAC BTK degraders.
  • Potent BTK inhibitor
  • Reversible Ibrutinib derivative
  • Useful as a BTK ligand for PROTAC synthesis
  • Effective against both WT BTK and C481S BTK
Catalog No. 50-003-38989 Supplier Medchemexpress LLC Supplier No. HY13098310MM/1ML
N-piperidine Ibrutinib hydrochloride is a reversible Ibrutinib derivative and a potent BTK inhibitor. It demonstrates high efficacy with IC50s of 51.0 nM for WT BTK and 30.7 nM for C481S BTK. This compound serves as a valuable BTK ligand in the synthesis of various PROTACs, including potent PROTAC BTK degraders.
  • Potent BTK inhibitor
  • Reversible Ibrutinib derivative
  • Useful as a BTK ligand for PROTAC synthesis
  • Effective against both WT BTK and C481S BTK