Learn More
Medchemexpress LLC N-piperidine Ibrutinib (hydrochloride) | 2231747-18-3 | 98.1% | 25 MG
SDP
Catalog No. 5000341429
Encompass_Preferred
N-piperidine Ibrutinib hydrochloride is a reversible Ibrutinib derivative and a potent BTK inhibitor. It can be used as a BTK ligand in the synthesis of a series of PROTACs.
  • Reversible Ibrutinib derivative
  • Potent BTK inhibitor with IC50s of 51.0 nM for WT BTK and 30.7 nM for C481S BTK
  • Used as a BTK ligand in the synthesis of PROTACs
  • Solid appearance, white to yellow color
  • For research use only
Catalog No. 50-003-41429 Supplier Medchemexpress LLC Supplier No. 25MGHY130983
N-piperidine Ibrutinib hydrochloride is a reversible Ibrutinib derivative and a potent BTK inhibitor. It can be used as a BTK ligand in the synthesis of a series of PROTACs.
  • Reversible Ibrutinib derivative
  • Potent BTK inhibitor with IC50s of 51.0 nM for WT BTK and 30.7 nM for C481S BTK
  • Used as a BTK ligand in the synthesis of PROTACs
  • Solid appearance, white to yellow color
  • For research use only