Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-piperidine ibrutinib hydrochloride | 2231747-18-3 | 98.1% | 422.91 g/mol | C22H23ClN6O | 5 MG
SDP

Catalog No. 5000262914
Encompass_Preferred

N-piperidine ibrutinib hydrochloride is a research-grade small molecule inhibitor of Bruton's tyrosine kinase (BTK) used in biochemical and pharmacological research, including PROTAC applications. It is supplied as a white-to-yellow solid with characterized purity and physicochemical properties for in vitro and in vivo formulation studies.

  • High purity (98.06%) solid suitable for biochemical assays.
  • Molecular weight 422.91 g/mol; formula C22H23ClN6O.
  • Soluble in DMSO and water; specific preparation protocols recommended.
  • Storage recommendations provided for solid and solution stability.
  • Intended for BTK-targeted studies and PROTAC research.
  • Available in small research quantities and DMSO solution formats.

Catalog No. 50-002-62914 Supplier Medchemexpress LLC Supplier No. HY1309835MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-piperidine ibrutinib hydrochloride is a research-grade small molecule inhibitor of Bruton's tyrosine kinase (BTK) used in biochemical and pharmacological research, including PROTAC applications. It is supplied as a white-to-yellow solid with characterized purity and physicochemical properties for in vitro and in vivo formulation studies.

  • High purity (98.06%) solid suitable for biochemical assays.
  • Molecular weight 422.91 g/mol; formula C22H23ClN6O.
  • Soluble in DMSO and water; specific preparation protocols recommended.
  • Storage recommendations provided for solid and solution stability.
  • Intended for BTK-targeted studies and PROTAC research.
  • Available in small research quantities and DMSO solution formats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.