Learn More
Medchemexpress LLC N-piperidine ibrutinib hydrochloride | 2231747-18-3 | 98.1% | C22H23ClN6O | 10MG
SDP
Catalog No. 5000258783
Encompass_Preferred
N-piperidine ibrutinib hydrochloride is a hydrochloride salt derivative used in research as a BTK ligand and synthetic intermediate for targeted degraders. It is supplied as a high-purity solid and as a ready-to-use DMSO solution, and is characterized for potency against wild-type and mutant BTK.
  • Potent BTK activity with low-nanomolar IC50s for WT and C481S variants.
  • High purity suitable for research applications.
  • Available as solid powder and 10 mM solution in DMSO.
  • White to yellow solid appearance for easy handling.
  • Useful as a ligand in PROTAC synthesis and other chemical biology workflows.
Catalog No. 50-002-58783 Supplier Medchemexpress LLC Supplier No. HY13098310MG
N-piperidine ibrutinib hydrochloride is a hydrochloride salt derivative used in research as a BTK ligand and synthetic intermediate for targeted degraders. It is supplied as a high-purity solid and as a ready-to-use DMSO solution, and is characterized for potency against wild-type and mutant BTK.
  • Potent BTK activity with low-nanomolar IC50s for WT and C481S variants.
  • High purity suitable for research applications.
  • Available as solid powder and 10 mM solution in DMSO.
  • White to yellow solid appearance for easy handling.
  • Useful as a ligand in PROTAC synthesis and other chemical biology workflows.