Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(Propargyl-PEG4)-biocytin | 2055042-71-0 | 98.0% | C28H46N4O9S | 100 MG
SDP

Catalog No. 5000353399
Encompass_Preferred

N-(Propargyl-PEG4)-biocytin is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This product is intended for research use only.

  • Molecular weight: 614.75
  • Purity: 98.0%
  • Appearance: Solid
  • Color: Light yellow to light brown
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)

Catalog No. 50-003-53399 Supplier Medchemexpress LLC Supplier No. HY140926100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(Propargyl-PEG4)-biocytin is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This product is intended for research use only.

  • Molecular weight: 614.75
  • Purity: 98.0%
  • Appearance: Solid
  • Color: Light yellow to light brown
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.