Learn More
Medchemexpress LLC N-(Propargyl-PEG4)-biocytin | 2055042-71-0 | 98.0% | C28H46N4O9S | 250 MG
SDP
Catalog No. 5000352748
Encompass_Preferred
N-(Propargyl-PEG4)-biocytin is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs consist of two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other the protein of interest, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports targeted protein degradation
Catalog No. 50-003-52748 Supplier Medchemexpress LLC Supplier No. 250MGHY140926
N-(Propargyl-PEG4)-biocytin is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs consist of two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other the protein of interest, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports targeted protein degradation