Learn More
Medchemexpress LLC N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) | 2112732-01-9 | 519.58 | 25 MG
SDP
Catalog No. 5000351405
Encompass_Preferred
N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs and acts as a click chemistry reagent. This linker contains an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess azide groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-51405 Supplier Medchemexpress LLC Supplier No. 25MGHY140095
N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs and acts as a click chemistry reagent. This linker contains an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess azide groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups