Learn More
Medchemexpress LLC N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) | 2112732-01-9 | 519.58 | 50 MG
SDP
Catalog No. 5000349803
Encompass_Preferred
N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (Cuaac) with azide groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49803 Supplier Medchemexpress LLC Supplier No. HY14009550MG
N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (Cuaac) with azide groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins