Learn More
Medchemexpress LLC N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) | 2112732-01-9 | > 96% | 519.58 | 250 MG
SDP
Catalog No. 5000351135
Encompass_Preferred
N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Allows copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-51135 Supplier Medchemexpress LLC Supplier No. 250MGHY140095
N-(Propargyl-PEG4-carbonyl)-N-bis(PEG1-methyl ester) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Allows copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups