Learn More
Medchemexpress LLC N-propargylglycine | 58160-95-5 | 99.7% | C5H7NO2 | 10MG
SDP
Catalog No. 5000261368
Encompass_Preferred
N-propargylglycine is an orally active, irreversible inhibitor of proline dehydrogenase (PRODH) used in biochemical and pharmacological research. It exhibits reported antitumor activity and is applied in studies of mitochondrial metabolism and neurodegenerative disease mechanisms. Available as a solid and as a 10 mM solution in DMSO, it has formula C5H7NO2 and reported purity 99.7%.
  • Irreversible inhibitor of proline dehydrogenase (PRODH).
  • Orally active small molecule suitable for in vivo and in vitro studies.
  • Reported antitumor activity supports oncology research.
  • Useful for studies of mitochondrial metabolism and neurodegenerative disease mechanisms.
  • Offered in solid form and as a 10 mM solution in DMSO for convenience.
Catalog No. 50-002-61368 Supplier Medchemexpress LLC Supplier No. HY14203510MG
N-propargylglycine is an orally active, irreversible inhibitor of proline dehydrogenase (PRODH) used in biochemical and pharmacological research. It exhibits reported antitumor activity and is applied in studies of mitochondrial metabolism and neurodegenerative disease mechanisms. Available as a solid and as a 10 mM solution in DMSO, it has formula C5H7NO2 and reported purity 99.7%.
  • Irreversible inhibitor of proline dehydrogenase (PRODH).
  • Orally active small molecule suitable for in vivo and in vitro studies.
  • Reported antitumor activity supports oncology research.
  • Useful for studies of mitochondrial metabolism and neurodegenerative disease mechanisms.
  • Offered in solid form and as a 10 mM solution in DMSO for convenience.