Learn More
Medchemexpress LLC N-propargylglycine | 58160-95-5 | MFCD11168312 | 99.7% | 113.11 | C5H7NO2 | 5 MG
SDP
Catalog No. 5000221359
Encompass_Preferred
N-propargylglycine is an orally active, irreversible inhibitor of proline dehydrogenase (PRODH) used in research on mitochondrial metabolism and neurodegenerative diseases. It has reported antitumor activity and is supplied as an off-white to light-yellow solid for laboratory research.
  • Orally active, irreversible PRODH inhibitor.
  • Applicable to studies of mitochondrial metabolism and neurodegeneration.
  • Reported antitumor activity supports cancer research use.
  • High purity (99.7%) for reproducible experimental results.
  • Solid form facilitates storage and handling in the laboratory.
  • Available in small package sizes suitable for screening and pilot studies.
Catalog No. 50-002-21359 Supplier Medchemexpress LLC Supplier No. HY1420355MG
N-propargylglycine is an orally active, irreversible inhibitor of proline dehydrogenase (PRODH) used in research on mitochondrial metabolism and neurodegenerative diseases. It has reported antitumor activity and is supplied as an off-white to light-yellow solid for laboratory research.
  • Orally active, irreversible PRODH inhibitor.
  • Applicable to studies of mitochondrial metabolism and neurodegeneration.
  • Reported antitumor activity supports cancer research use.
  • High purity (99.7%) for reproducible experimental results.
  • Solid form facilitates storage and handling in the laboratory.
  • Available in small package sizes suitable for screening and pilot studies.