Learn More
Medchemexpress LLC N-ε-propargyloxycarbonyl-L-lysine hydrochloride | 1428330-91-9 | 99.8% | 1 ML
SDP
Catalog No. 5000341996
Encompass_Preferred
N-ε-propargyloxycarbonyl-L-lysine hydrochloride is a lysine-based unnatural amino acid (UAA) primarily used for bio-conjugation and click chemistry applications.
  • Lysine-based unnatural amino acid
  • Used for bio-conjugation of fluorescent probes in diverse organisms
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • For research use only
Catalog No. 50-003-41996 Supplier Medchemexpress LLC Supplier No. HY12867610MM/1ML
N-ε-propargyloxycarbonyl-L-lysine hydrochloride is a lysine-based unnatural amino acid (UAA) primarily used for bio-conjugation and click chemistry applications.
  • Lysine-based unnatural amino acid
  • Used for bio-conjugation of fluorescent probes in diverse organisms
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • For research use only