Learn More
Sigma Aldrich Fine Chemicals Biosciences n-Propionyl coenzyme A lit5MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P53975MG
Encompass
Propionyl-CoA is obtained as an end product of isoleucine valine and methionine catabolism. It is an essential component for the methylaspartate cycle. Branched-chain amino acids and cholesterol also give rise to propionyl-CoA. Propionyl coenzyme A (CoA) is the coenzyme A derivative of propionic acid. Propionyl CoA is formed during the -oxidation of odd-chain fatty acids.
Catalog No. 50-290-2133