Learn More
Cayman Chemical N-Propionylglycine, Purity: ≥95%, CAS: 21709-90-0, 100 mg
SDP
Catalog No. 502947303
Encompass_Preferred
A glycine-conjugated form of propionic acid; urinary levels are increased in patients with propionic acidemia
Catalog No. 50-294-7303 Supplier Cayman Chemical Supplier No. 34377100 MG
A glycine-conjugated form of propionic acid; urinary levels are increased in patients with propionic acidemia