Learn More
Cayman Chemical N-PropIonylglycIn 50mg
SDP
Catalog No. 502950995
Encompass_Preferred
A glycine-conjugated form of propionic acid; urinary levels are increased in patients with propionic acidemia
Catalog No. 50-295-0995 Supplier Cayman Chemical Supplier No. 3437750 MG
A glycine-conjugated form of propionic acid; urinary levels are increased in patients with propionic acidemia