Learn More
n-Propyl 4-hydroxybenzoate, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
n-Propyl 4-hydroxybenzoate is used as antimicrobial preservatives in pharmaceuticals, foods, cosmetics and shampoos. It is widely used as preservatives in personal care products like creams, lotions and bath products. Further, it is used as a food additive. Its derivative, sodium propyl p-hydroxybenzoate is used as an anti-fungal preservation agent.
Solubility
Soluble in propylene glycol, ethanol, ether, acetone, methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and alkali metals.
Specifications
Specifications
| Melting Point | 95°C to 99°C |
| Flash Point | 180°C (356°F) |
| Quantity | 5000 g |
| Beilstein | 1103245 |
| Merck Index | 14,7866 |
| Solubility Information | Soluble in propylene glycol,ethanol,ether,acetone,methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water. |
| Formula Weight | 180.2 |
| Percent Purity | ≥99% |
| Chemical Name or Material | n-Propyl 4-hydroxybenzoate |
Safety and Handling
| Hazard Category | H315-H320-H373 |
| Precautionary Statement | P262-P264b-P280-P302+P352-P305+P351+P338-P314-P332+P313-P362-P501c |
| EINECSNumber | 202-307-7 |
| RTECSNumber | DH2800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.