Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp n-Propyl 4-hydroxybenzoate 1KG
SDP

Catalog No. 502230299
Encompass_Preferred

Purity:>99%
Store at: Room Temperature
CAS No. 94-13-3

Catalog No. 50-223-0299 Supplier Research Products International Corp Supplier No. P503201000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purity:>99%
Store at: Room Temperature
CAS No. 94-13-3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.