Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Propyl Acetate, Reagent Grade, Reagents

Catalog No. 01338131
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-131 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-131 Supplier Reagents Holdings Llc Supplier No. C11210104C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Propyl Acetate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym Propyl Ethanoate, Acetic Acid Propyl Ester
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

Melting Point -93°C
Color Colorless
Boiling Point 101.5°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.886
Grade Reagent
Physical Form Liquid
Chemical Name or Material N-Propyl Acetate, Reagent Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.