Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyl decanoate, 98%

Catalog No. AAB2515918
Click to view available options
Quantity:
50 g
250 g
1000 g

Catalog No. AAB2515918

Supplier: Thermo Scientific Chemicals B2515918

Only null left
Add to Cart

CAS: 30673-60-0 | C13H26O2 | 214.349 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30673-60-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00056329
InChI Key OVFMRFMJVFDSAA-UHFFFAOYSA-N
Synonym n-propyl decanoate, decanoic acid, propyl ester, propyl caprate, capric acid propyl ester, unii-s0he9grs1w, decanoic acid, propylester, s0he9grs1w, propyl caprate natural, propyl caprate decanoate, n-propyldecanoate
PubChem CID 121739
ChEBI CHEBI:86973
IUPAC Name propyl decanoate
SMILES CCCCCCCCCC(=O)OCCC

Specifications

Density 0.86
Boiling Point 254°C
Flash Point 115°C (239°F)
Quantity 50 g
Formula Weight 214.35
Percent Purity 98%
Chemical Name or Material n-Propyl decanoate
EINECSNumber 250-283-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.