Learn More
Cayman Chemical N-propyl-L-ArgInIn 10mg
SDP
Catalog No. 502896464
Encompass_Preferred
A potent and selective competitive inhibitor of nNOS (Ki = 57 nM; bovine); exhibits 3,000-fold and 150-fold selectivity for the neuronal isoform versus the inducible (murine; Ki = 180 µM) and endothelial (bovine; Ki = 8.5 µM) isoforms of NOS, respectively
Catalog No. 50-289-6464 Supplier Cayman Chemical Supplier No. 8058710 MG
A potent and selective competitive inhibitor of nNOS (Ki = 57 nM; bovine); exhibits 3,000-fold and 150-fold selectivity for the neuronal isoform versus the inducible (murine; Ki = 180 µM) and endothelial (bovine; Ki = 8.5 µM) isoforms of NOS, respectively