Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyltriacetoxysilane, 98%, Thermo Scientific™

Catalog No. AAH60679MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60679MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH60679MD Supplier Thermo Scientific Chemicals Supplier No. H60679MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17865-07-5
Molecular Formula C9H16O6Si
Molecular Weight (g/mol) 248.306
MDL Number MFCD00086768
InChI Key DKGZKEKMWBGTIB-UHFFFAOYSA-N
Synonym propylsilanetriyl triacetate, propyltriacetoxysilane, n-propyltriacetoxysilane, silanetriol, propyl-, triacetate, silanetriol, 1-propyl-, 1,1,1-triacetate, bis acetyloxy propyl silyl acetate, triacetoxypropylsilane, propyl-silanetriotriacetate, propylsilanetriol triacetate, diacetyloxy propyl silyl acetate
PubChem CID 87339
IUPAC Name [diacetyloxy(propyl)silyl] acetate
SMILES CCC[Si](OC(=O)C)(OC(=O)C)OC(=O)C

Specifications

Quantity 250 mg
Formula Weight 248.31
Percent Purity 98%
Chemical Name or Material n-Propyltriacetoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.