Learn More
Medchemexpress LLC N-(pyridin-3-yl)thiophene-2-carboxamide | 62289-81-0 | MFCD00469725 | 99.7% | 204.25 g/mol | C10H8N2OS | 25MG
SDP
Catalog No. 5000238136
Encompass_Preferred
SW106065 (N-(pyridin-3-yl)thiophene-2-carboxamide; CAS 62289-81-0) is a small-molecule research compound that induces apoptosis and inhibits proliferation in malignant peripheral nerve sheath tumor (MPNST) cell lines in vitro and reduces tumor growth in mouse models. Supplied in high-purity solid and solution formats for laboratory research use only.
  • High purity suitable for research applications.
  • Available in multiple sizes and solution formats for experimental flexibility.
  • Shown to induce apoptosis and inhibit MPNST cell proliferation in vitro.
  • Demonstrated in vivo activity in mouse tumor models.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-38136 Supplier Medchemexpress LLC Supplier No. HY12477825MG
SW106065 (N-(pyridin-3-yl)thiophene-2-carboxamide; CAS 62289-81-0) is a small-molecule research compound that induces apoptosis and inhibits proliferation in malignant peripheral nerve sheath tumor (MPNST) cell lines in vitro and reduces tumor growth in mouse models. Supplied in high-purity solid and solution formats for laboratory research use only.
  • High purity suitable for research applications.
  • Available in multiple sizes and solution formats for experimental flexibility.
  • Shown to induce apoptosis and inhibit MPNST cell proliferation in vitro.
  • Demonstrated in vivo activity in mouse tumor models.
  • Stable under recommended storage conditions for long-term use.