Learn More
Medchemexpress LLC N,S-Bis-Fmoc-Glutathione | 149438-56-2 | 99.9% | 751.80 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP4277
Encompass_Preferred
N,S-Bis-Fmoc-Glutathione is recognized as a potent glyoxalase II inhibitor, demonstrating a Ki value of 0.75 μM and an IC50 of 2.5 μM.
  • Potent glyoxalase II inhibitor
  • Ki value: 0.75 μM
  • IC50 value: 2.5 μM
  • Appears as a solid
  • Color is white to off-white
Catalog No. 50-004-00265