Learn More
Medchemexpress LLC N,S-Bis-Fmoc-Glutathione (TFA) | 97.1% | 865.82 | 25 MG
SDP
Catalog No. 5000396583
Encompass_Preferred
N,S-Bis-Fmoc-Glutathione (TFA) is a chemical compound supplied as a white to off-white solid, intended for laboratory research and manufacturing. It offers high purity and stability when stored under recommended conditions.
  • High purity (97.1% by LCMS)
  • Available as a white to off-white solid
  • Stable under recommended storage conditions
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
  • Recommended storage for solutions: -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-96583 Supplier Medchemexpress LLC Supplier No. 25MGHYP4277A
N,S-Bis-Fmoc-Glutathione (TFA) is a chemical compound supplied as a white to off-white solid, intended for laboratory research and manufacturing. It offers high purity and stability when stored under recommended conditions.
  • High purity (97.1% by LCMS)
  • Available as a white to off-white solid
  • Stable under recommended storage conditions
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
  • Recommended storage for solutions: -80°C for 6 months or -20°C for 1 month