Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Selectride™

Catalog No. 213403100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-340-3100ML 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 21-340-3100ML Supplier Sigma Aldrich Supplier No. 213403100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reducing agentReactant for:Conjugate hydride reduction-initiated tandem cyclization reactionsSereoselective nucleophilic addition reactionsSelectride is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 207500-00-3
Molecular Formula Pd
Linear Formula Pd
MDL Number MFCD03457879
Quantity 100 g
Synonym Sodium tri-sec-butylborohydride solution
Molecular Weight (g/mol) 106.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.