Learn More
Medchemexpress LLC N-succinimidyl-3-(2-pyridyldithio)butyrate | 107348-47-0 | MFCD28398150 | 98.3% | 326.4 g/mol | C13H14N2O4S2 | 1 G
SDP
Catalog No. 5000203079
Encompass_Preferred
PPC-NHS ester (N-succinimidyl-3-(2-pyridyldithio)butyrate, CAS 107348-47-0) is an NHS-activated cleavable linker reagent used for conjugating small molecules to proteins and antibodies. It reacts with primary amines to form stable amide bonds and contains a pyridyldisulfide moiety that enables controlled cleavage under reducing conditions, making it suitable for antibody-drug conjugate synthesis and general bioconjugation.
  • Reacts with primary amines to form stable amide bonds.
  • Contains a pyridyldisulfide cleavable linkage for controlled release under reducing conditions.
  • High purity suitable for conjugation workflows (≈98.3%).
  • Compatible with common aqueous and organic conjugation conditions.
Catalog No. 50-002-03079 Supplier Medchemexpress LLC Supplier No. HYW0710061G
PPC-NHS ester (N-succinimidyl-3-(2-pyridyldithio)butyrate, CAS 107348-47-0) is an NHS-activated cleavable linker reagent used for conjugating small molecules to proteins and antibodies. It reacts with primary amines to form stable amide bonds and contains a pyridyldisulfide moiety that enables controlled cleavage under reducing conditions, making it suitable for antibody-drug conjugate synthesis and general bioconjugation.
  • Reacts with primary amines to form stable amide bonds.
  • Contains a pyridyldisulfide cleavable linkage for controlled release under reducing conditions.
  • High purity suitable for conjugation workflows (≈98.3%).
  • Compatible with common aqueous and organic conjugation conditions.