Learn More
Medchemexpress LLC N-succinimidyl 3-(acetylthio)propionate | 84271-78-3 | MFCD00133023 | >95.0% | 245.25 g/mol | C9H11NO5S | 5 G
SDP
Catalog No. 5000303235
Encompass_Preferred
3-(Acetylthio)propionic acid N-succinimidyl ester is an N-hydroxysuccinimide (NHS) activated ester used to introduce a protected thiol-containing propionate linker onto amine-functionalized molecules. It is commonly used in bioconjugation and linker synthesis applications where an amine-reactive NHS ester and a masked thiol are required. Typical properties include formula C9H11NO5S and molecular weight 245.25 g/mol.
  • Provides an NHS-activated carboxyl for efficient amide bond formation.
  • Delivers an acetyl-protected thiol for controlled deprotection and subsequent conjugation.
  • Suitable for bioconjugation, linker assembly, and small-molecule functionalization workflows.
  • Offered in multiple packaging sizes to support research-scale needs.
  • Reported supplier purities are typically ≥95%; verify lot-specific certificate of analysis prior to use.
Catalog No. 50-003-03235 Supplier Medchemexpress LLC Supplier No. HYW0139765G
3-(Acetylthio)propionic acid N-succinimidyl ester is an N-hydroxysuccinimide (NHS) activated ester used to introduce a protected thiol-containing propionate linker onto amine-functionalized molecules. It is commonly used in bioconjugation and linker synthesis applications where an amine-reactive NHS ester and a masked thiol are required. Typical properties include formula C9H11NO5S and molecular weight 245.25 g/mol.
  • Provides an NHS-activated carboxyl for efficient amide bond formation.
  • Delivers an acetyl-protected thiol for controlled deprotection and subsequent conjugation.
  • Suitable for bioconjugation, linker assembly, and small-molecule functionalization workflows.
  • Offered in multiple packaging sizes to support research-scale needs.
  • Reported supplier purities are typically ≥95%; verify lot-specific certificate of analysis prior to use.