Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 3-(Acetylthio)propionate 95.0+%, TCI America™
SDP

Catalog No. S0859100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0859100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0859100MG Supplier TCI America Supplier No. S0859100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84271-78-3
Molecular Formula C9H11NO5S
Molecular Weight (g/mol) 245.249
MDL Number MFCD00133023
InChI Key ZRTJVRDXVSDKPX-UHFFFAOYSA-N
Synonym 3-(Acetylthio)propionic Acid N-Succinimidyl Ester, SATP
PubChem CID 4110341
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-acetylsulfanylpropanoate
SMILES CC(=O)SCCC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 66°C
Color White
Quantity 100 mg
Formula Weight 245.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 3-(Acetylthio)propionate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.