Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 3-(Diphenylphosphino)propionate 98.0+%, TCI America™
SDP

Catalog No. S08165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08165G 5 g
S08161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08165G Supplier TCI America Supplier No. S08165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 170278-50-9
Molecular Formula C19H18NO4P
Molecular Weight (g/mol) 355.33
InChI Key QRRPSWGREIRROT-UHFFFAOYSA-N
Synonym 3-(Diphenylphosphino)propionic Acid N-Succinimidyl Ester
PubChem CID 42604311
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-diphenylphosphanylpropanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCP(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Color White
Quantity 5 g
Formula Weight 355.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 3-(Diphenylphosphino)propionate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.