Learn More
Medchemexpress LLC N-succinimidyl 3-maleimidopropionate | 55750-62-4 | MFCD00043141 | 99.9% | 266.21 g/mol | C11H10N2O6 | 25 G
SDP
Catalog No. 5000204077
Encompass_Preferred
BMPS (N-succinimidyl 3-maleimidopropionate) is a heterobifunctional maleimide-NHS ester used as a noncleavable linker or crosslinker for amine-to-sulfhydryl conjugation in antibody-drug conjugate (ADC) synthesis and other bioconjugation chemistries.
  • Heterobifunctional maleimide-NHS ester for amine-to-sulfhydryl conjugation.
  • Noncleavable linker suitable for ADC and protein crosslinking applications.
  • High purity (≈99.9%) solid that facilitates stable conjugate formation.
  • Formula C11H10N2O6 and molecular weight 266.21 g/mol.
  • Intended for research use only; handle and store according to SDS instructions.
Catalog No. 50-002-04077 Supplier Medchemexpress LLC Supplier No. HY4214625G
BMPS (N-succinimidyl 3-maleimidopropionate) is a heterobifunctional maleimide-NHS ester used as a noncleavable linker or crosslinker for amine-to-sulfhydryl conjugation in antibody-drug conjugate (ADC) synthesis and other bioconjugation chemistries.
  • Heterobifunctional maleimide-NHS ester for amine-to-sulfhydryl conjugation.
  • Noncleavable linker suitable for ADC and protein crosslinking applications.
  • High purity (≈99.9%) solid that facilitates stable conjugate formation.
  • Formula C11H10N2O6 and molecular weight 266.21 g/mol.
  • Intended for research use only; handle and store according to SDS instructions.