Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 3-(propargyloxy)propionate

Catalog No. AAJ64496MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64496MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64496MC Supplier Thermo Scientific Chemicals Supplier No. J64496MC
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1174157-65-3 | C10H11NO5 | 225.20 g/mol

N-Succinimidyl 3-(propargyloxy)propionate is an acetylene-containing reagent with NHS ester functionality for reaction with amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 3-(propargyloxy)propionate is an acetylene-containing reagent with NHS ester functionality for reaction with amines.

Solubility
Slightly soluble in water.

Notes
Keep container tightly
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1174157-65-3
Molecular Formula C10H11NO5
Molecular Weight (g/mol) 225.20
MDL Number MFCD13184973
InChI Key WKIKHHMUNOVQLD-UHFFFAOYSA-N
Synonym n-succinimidyl 3-propargyloxy propionate, propargyl-nhs, ambotzpeg1935, acetylene-peg-nhs ester, propargyl-peg1-nhs ester, propargyl-succinimidyl-ester, propargyl-n-hydroxysuccinimidyl ester, 2,5-dioxopyrrolidin-1-yl 3-prop-2-ynyloxy propanoate, 2,5-dioxopyrrolidin-1-yl 3-prop-2-yn-1-yloxy propanoate
PubChem CID 51340986
IUPAC Name 2,5-dioxopyrrolidin-1-yl 3-(prop-2-yn-1-yloxy)propanoate
SMILES O=C(CCOCC#C)ON1C(=O)CCC1=O

Specifications

Color Gray
Quantity 100 mg
Solubility Information Slightly soluble in water.
Formula Weight 225.2
Physical Form Solid
Chemical Name or Material N-Succinimidyl 3-(propargyloxy)propionate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.