Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Succinimidyl 4-[4-maleimidophenyl]butyrate | 79886-55-8 | 98.0% | 356.33 | 250 MG
SDP

Catalog No. 5000453332
Encompass_Preferred

N-Succinimidyl 4-[4-maleimidophenyl]butyrate is a chemical compound that functions as a crosslinking agent. It is intended for research use only and has not been fully validated for medical applications.

  • Molecular formula: C18H16N2O6
  • Target: DNA alkylator/crosslinker
  • Pathway: Cell cycle/DNA damage
  • Storage conditions: -20°C, stored under nitrogen, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month
  • Solubility (in vitro): DMSO: 100 mg/mL (280.64 mM)
  • Appearance: White to off-white solid
  • Melting point: 130-131 °C
  • Boiling point: 551.1±60.0 °C at 760 mmHg

Catalog No. 50-004-53332 Supplier Medchemexpress LLC Supplier No. 250MGHYW011123
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Succinimidyl 4-[4-maleimidophenyl]butyrate is a chemical compound that functions as a crosslinking agent. It is intended for research use only and has not been fully validated for medical applications.

  • Molecular formula: C18H16N2O6
  • Target: DNA alkylator/crosslinker
  • Pathway: Cell cycle/DNA damage
  • Storage conditions: -20°C, stored under nitrogen, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month
  • Solubility (in vitro): DMSO: 100 mg/mL (280.64 mM)
  • Appearance: White to off-white solid
  • Melting point: 130-131 °C
  • Boiling point: 551.1±60.0 °C at 760 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.