Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate

Catalog No. AAJ64902MC
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64902MC 100 mg
AAJ64902MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64902MC Supplier Thermo Scientific Chemicals Supplier No. J64902MC
Add to Cart

C18H27NO9

N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate is a labeling reagent with amine reactivity as the activated NHS ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate is a labeling reagent with amine reactivity as the activated NHS ester.

Solubility
Slightly soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C18H27NO9
MDL Number MFCD22380743
Synonym Acetylene-PEG5-NHS ester

Specifications

Color Orange to Yellow
Quantity 100 mg
Solubility Information Slightly soluble in dimethyl sulfoxide and dimethyl formamide.
Formula Weight 401.41
Physical Form Viscous Liquid
Chemical Name or Material N-Succinimidyl 4,7,10,13,16-pentaoxanonadec-18-ynoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.