Learn More
Medchemexpress LLC N-succinimidyl 4-(dimethylamino)benzoate | 58068-85-2 | MFCD16875522 | 99.0% | 262.26 g/mol | C13H14N2O4 | 10 MG
SDP
Catalog No. 5000227467
Encompass_Preferred
DMABA NHS ester is an N-hydroxysuccinimide (NHS) ester reagent used to derivatize primary amine groups of phosphatidylethanolamine (PE) lipids, enabling selective detection by mass spectrometry. It is supplied as a white to off-white solid with high purity and recommended cold storage to preserve activity.
  • Enables selective labeling of PE lipids for precursor ion scanning.
  • Improves detection sensitivity in electrospray tandem mass spectrometry.
  • High purity supports reliable analytical results.
  • Stable as supplied with recommended low-temperature, moisture-free storage.
  • Compatible with isotope-labeled DMABA analogs for comparative analyses.
Catalog No. 50-002-27467 Supplier Medchemexpress LLC Supplier No. HY11843110MG
DMABA NHS ester is an N-hydroxysuccinimide (NHS) ester reagent used to derivatize primary amine groups of phosphatidylethanolamine (PE) lipids, enabling selective detection by mass spectrometry. It is supplied as a white to off-white solid with high purity and recommended cold storage to preserve activity.
  • Enables selective labeling of PE lipids for precursor ion scanning.
  • Improves detection sensitivity in electrospray tandem mass spectrometry.
  • High purity supports reliable analytical results.
  • Stable as supplied with recommended low-temperature, moisture-free storage.
  • Compatible with isotope-labeled DMABA analogs for comparative analyses.