Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 6-Maleimidohexanoate 98.0+%, TCI America™
SDP

Catalog No. S04281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04281G Supplier TCI America Supplier No. S04281G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 55750-63-5
Melting Point 71°C
Color White-Yellow
Molecular Formula C14H16N2O6
Quantity 1 g
MDL Number MFCD00043043
Synonym n-succinimidyl 6-maleimidohexanoate, emcs, 6-maleimidohexanoic acid n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoate, n-succinimidyl 6-maleimidocaproate, n-6-maleimidocaproyloxy succinimide, 2,5-dioxopyrrolidin-1-yl 6-2,5-dioxopyrrol-1-yl hexanoate, nmicos, 1h-pyrrole-2,5-dione, 1-6-2,5-dioxo-1-pyrrolidinyl oxy-6-oxohexyl
InChI Key VLARLSIGSPVYHX-UHFFFAOYSA-N
SMILES O=C(CCCCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O
IUPAC Name 2,5-dioxopyrrolidin-1-yl 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate
Molecular Weight (g/mol) 308.29
PubChem CID 5091655
Formula Weight 308.29
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 6-Maleimidohexanoate [Cross-linking Reagent]
Show More Show Less
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.