Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl Acetate 98.0+%, TCI America™
SDP

Catalog No. S087825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S087825G 25 g
S08785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S087825G Supplier TCI America Supplier No. S087825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14464-29-0
Molecular Formula C6H7NO4
Molecular Weight (g/mol) 157.125
MDL Number MFCD00037916
InChI Key SIFCHNIAAPMMKG-UHFFFAOYSA-N
Synonym Acetic Acid N-Succinimidyl Ester, N-Acetoxysuccinimide
PubChem CID 84460
IUPAC Name (2,5-dioxopyrrolidin-1-yl) acetate
SMILES CC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 157.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl Acetate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.