Learn More
Medchemexpress LLC N-succinimidyl iodoacetate | 39028-27-8 | MFCD00058451 | >95.0% | 283.02 | C6H6INO4 | 250 MG
SDP
Catalog No. 5000203187
Encompass_Preferred
SIA Crosslinker (N-succinimidyl iodoacetate) is a short, heterobifunctional crosslinker used to form stable amine-to-sulfhydryl conjugates. It serves as a non-cleavable ADC linker and is used for protein-protein and protein-small molecule conjugation.
  • Amine- and sulfhydryl-reactive (NHS ester and iodoacetyl).
  • Very short spacer arm (~1.5 Å) for minimal linkage length.
  • Molecular weight 283.02, formula C6H6INO4, CAS 39028-27-8.
  • Used for antibody-drug conjugates and general protein conjugation.
  • Available in common laboratory package sizes (100 mg, 250 mg, 500 mg).
Catalog No. 50-002-03187 Supplier Medchemexpress LLC Supplier No. HYW011541250MG
SIA Crosslinker (N-succinimidyl iodoacetate) is a short, heterobifunctional crosslinker used to form stable amine-to-sulfhydryl conjugates. It serves as a non-cleavable ADC linker and is used for protein-protein and protein-small molecule conjugation.
  • Amine- and sulfhydryl-reactive (NHS ester and iodoacetyl).
  • Very short spacer arm (~1.5 Å) for minimal linkage length.
  • Molecular weight 283.02, formula C6H6INO4, CAS 39028-27-8.
  • Used for antibody-drug conjugates and general protein conjugation.
  • Available in common laboratory package sizes (100 mg, 250 mg, 500 mg).