Learn More
AdipoGen N-succinimidyl propionate
SDP
Catalog No. 502561070
Encompass_Preferred
Chemical. CAS 30364-55-7. Formula C7H9NO4. MW 171.15. Synthetic. Biochemical for proteomics research use. Amine-specific protein-acylating agent. Used for derivatization of biogenic amines for hplc analysis. Recent stuides use this agent for derivatization of histone post-translational modifications. The compound has been used in several studies describing this well established reagent for the introduction of a tritium label into proteins or other biologicals. The retrograde transport of protein components of sciatic nerve axons has been demonstrated by direct labelling of the protein usingNSP in vivo.
Catalog No. 50-256-1070 Supplier AdipoGen Supplier No. CDXD0489M250
Chemical. CAS 30364-55-7. Formula C7H9NO4. MW 171.15. Synthetic. Biochemical for proteomics research use. Amine-specific protein-acylating agent. Used for derivatization of biogenic amines for hplc analysis. Recent stuides use this agent for derivatization of histone post-translational modifications. The compound has been used in several studies describing this well established reagent for the introduction of a tritium label into proteins or other biologicals. The retrograde transport of protein components of sciatic nerve axons has been demonstrated by direct labelling of the protein usingNSP in vivo.