Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl Trifluoroacetate 95.0+%, TCI America™
SDP

Catalog No. S09151G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09151G Supplier TCI America Supplier No. S09151G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5672-89-9
Molecular Formula C6H4F3NO4
Molecular Weight (g/mol) 211.096
MDL Number MFCD00595252
InChI Key NIANZCQUVKPGSF-UHFFFAOYSA-N
Synonym Trifluoroacetic Acid N-Succinimidyl Ester, N-(Trifluoroacetoxy)succinimide, TFA-NHS
PubChem CID 10867564
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2,2,2-trifluoroacetate
SMILES C1CC(=O)N(C1=O)OC(=O)C(F)(F)F

Specifications

Melting Point 75°C
Color White
Boiling Point 100°C
Quantity 1 g
Formula Weight 211.10
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.