Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013590 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3590 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3590 Supplier MilliporeSigma Supelco Supplier No. 2920850MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD03095505
Synonym N-Succinimidyl [tris(2,4,6-trimethoxyphenyl)phosphonio]acetate bromide; TMPP-Ac-OSu

Specifications

Melting Point 199°C to 205°C
CAS 226409-58-1
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C33H39 BrNO13 P
Quantity 50 mg
Formula Weight 768.54
Percent Purity ≥98.5% (HPLC); ≥98.5%
Grade For protein sequence analysis (by MALDI-MS)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.