Learn More
Medchemexpress LLC N-succinyl-Ala-Ala-Pro-Phe-7-amido-4-methylcoumarin | 88467-45-2 | MFCD00080246 | 99.5% | C34H39N5O9 | 10MG
SDP
Catalog No. 5000259659
Encompass_Preferred
Suc-AAPF-AMC is a fluorogenic peptide substrate used for quantitative measurement of α-chymotrypsin activity. Enzymatic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for kinetic and end-point assays in solution.
  • Generates a fluorescent AMC signal upon enzymatic cleavage.
  • Suitable for sensitive, quantitative chymotrypsin activity assays.
  • High reported purity (99.5%).
  • Offered as solid or as a DMSO stock solution for flexible assay formats.
  • Stable when stored sealed, away from moisture and light at recommended temperatures.
Catalog No. 50-002-59659 Supplier Medchemexpress LLC Supplier No. HY13781110MG
Suc-AAPF-AMC is a fluorogenic peptide substrate used for quantitative measurement of α-chymotrypsin activity. Enzymatic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal suitable for kinetic and end-point assays in solution.
  • Generates a fluorescent AMC signal upon enzymatic cleavage.
  • Suitable for sensitive, quantitative chymotrypsin activity assays.
  • High reported purity (99.5%).
  • Offered as solid or as a DMSO stock solution for flexible assay formats.
  • Stable when stored sealed, away from moisture and light at recommended temperatures.