Learn More
Bachem H-Val-His-Leu-Thr-Pro-OH
SDP
Supplier:  Bachem 4010555.0025
Encompass_Preferred
H-Val-His-Leu-Thr-Pro-OH, 93913-38-3, C26H43N7O7, Mr 565.67, 25mg. N-terminal pentapeptide of human hemoglobin beta (Hb A and and the pathological HbS, which differ in position 6). Cellulose membrane-bound VHLTP was used as ligand for the anti-HbA1c antibody.
Catalog No. 50-194-6947