Learn More
Medchemexpress LLC HY-P1170 1mg Medchemexpress, N-terminally acetylated Leu-enkephalin CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP11701MG
Encompass_Preferred
Medchemexpress, HY-P1170 1mg N-terminally acetylated Leu-enkephalin CAS: N-terminally acetylated Leu-enkephalin is the N-terminally acetylated form of Leu-enkephalin. Leu-enkephalin is a five amino acid endogenous peptide that acts as an agonist at opioid receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4159